Learn More
Octamethylferrocene, TRC
Product Code. 30137054
Change view
Click to view available options
Quantity:
100 mg
500 mg
1 g
Unit Size:
100mg
1g
500mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30137054 1 g 1g
17971515 500 mg 500mg
17922395 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30137054 Supplier Toronto Research Chemicals Supplier No. TRCO2360051G
 Restricted Item
Estimated Shipment: 27-08-2026
Add to basket
Add to basket
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 59568-28-4
Molecular Formula C18H26Fe
Molecular Weight (g/mol) 298.24
Synonym 1,1',2,2',3,3',4,4'-Octamethylferrocene, sym-Octamethylferrocene, Bis(tetramethylcyclopentadienyl)iron(II)
IUPAC Name 1,1′,2,2′,3,3′,4,4′-Octamethylferrocene
SMILES CC12=[CH]3[C-]4(C)C5(C)=C1(C)[Fe+2]36789254[CH]%10=C9(C)C8(C)=C7(C)[C-]%106C
Specifications
Quantity 1 g
InChI Formula InChI=1S/2C9H13.Fe/c2*1-6-5-7(2)9(4)8(6)3;/h2*5H,1-4H3;/q2*-1;+2
Formula Weight 298.14
Chemical Name or Material Octamethylferrocene