Learn More
Octadecyltriethoxysilane, 85%, BLD Pharm™
Specifications
Specifications
| CAS | 7399-00-0 |
| Molecular Formula | C24H52O3Si |
| MDL Number | MFCD00039875 |
| Quantity | 100 g |
| SMILES | CCCCCCCCCCCCCCCCCC[Si](OCC)(OCC)OCC |
| Molecular Weight (g/mol) | 416.75 |
| Percent Purity | 85% |
| Chemical Name or Material | Octadecyltriethoxysilane |