missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Octadecyl 3-(3,5-Di-tert-butyl-4-hydroxyphenyl)propionate, Mikromol™
Description
Octadecyl 3-(3,5-Di-tert-butyl-4-hydroxyphenyl)propionate
Specifications
Specifications
| Chemical Name or Material | Plastic additive 11 |
| CAS | 2082-79-3 |
| Analyte or Component Names | Plastic additive 11 |
| Shipping Condition | Room Temperature |
| Molecular Formula | C35 H62 O3 |
| Certifications/Compliance | ISO 9001 |
| Synonym | Plastic additive 11, 3,5-bis(1,1-Dimethylethyl)-4-hydroxybenzenepropanoic acid octadecyl ester, 3,5-Di-tert-butyl-4-hydroxyhydrocinnamic acid octadecyl ester, 2,6-Di-tert-butyl-4-(octadecanoxycarbonylethyl)phenol, 2,6-Di-tert-butyl-4-[[(octadecyloxy)carbonyl]ethyl]phenol, 3,5-Di-tert-butyl-4-hydroxyphenylpropionic acid octadecyl ester, 3-(3,5-Di-tert-butyl-4-hydroxyphenol)propionic acid octadecyl ester, 3-(3,5-Di-tert-butyl-4-hydroxyphenyl)propionic acid octadecyl ester, 4-Carboxyethyl)-2,6-di-tert-butyl-1-hydroxybenzene stearyl ester, ADK Stab AO 50, AO 4, AO 50, AT 76, Addiox 1016, Anox PP 18, Antioxidant 1076, Arenox A 76, Chemnox 1076, Chinox 1076, E 376, Hostanox 016, Hostanox M 016, Hostanox O 16, I 1076, IR 1076, IX 1076, Irganox 1068, Irganox 1076, Irganox 1076DWJ, Irganox 1906, Irganox 1976, Irganox I 1076, Irganox L 107, Irgastab PVC 76, JY 1076, KY 1076, L 107, M 4, Mark AO 50, Naugard 76, O 16, Octadecanyl β-(3,5-di-tert-butyl-4-hydroxyphenyl)propionate, Octadecyl 3,5-bis(1,1-dimethylethyl)-4-hydroxyphenylpropanoate, Octadecyl 3,5-di-tert-butyl-4-hydroxyhydrocinnamate, Octadecyl 3,5-di-tert-butyl-4-hydroxyphenylpropionate, Octadecyl 3,5-di-tert-butyl-4-oxyphenylpropionate, Octadecyl 3-(3,5-di-tert-butyl-4-hydroxyphenyl)propionate, Octadecyl 3-(3',5'-di-tert-butyl-4-hydroxyphenyl) propionate, Octadecyl 3-(4-hydroxy-3,5-di-tert-butylphenyl)propionate, Octadecyl 3-(4'-hydroxy-3',5'-di-tert-butylphenyl)propionate, Octadecyl 3-[3,5-Bis(1,1-dimethylethyl)-4-hydroxybenzene]propionate, Octadecyl β-(3,5-di-tert-butyl-4-hydroxyphenyl)propionate, Octadecyl β-(3,5-tert-butyl-4-hydroxyphenyl)propionate, Octadecyl β-(4-hydroxy-3,5-di-tert-butylphenyl)propionate, Octadecyl β-(4'-hydroxy-3',5'-di-tert-butylphenyl)propionate, Octadecyl-3-(3',5'-di-tert-butyl-4'-hydroxyphenyl)propenoate, Octadecyl-3-(3',5'-di-tert-butyl-4'-hydroxyphenyl)propionate, Quantox 1076, Ralox 530, Selosol F 773, Songnox 1076, Sonox 1076, Stearyl 3,5-di-tert-butyl-4-hydroxyhydrocinnamate, Stearyl 3-(3,5-di-tert-butyl-4-hydroxyphenyl)propionate, Stearyl β-(3,5-di-tert-butyl-4-hydroxyphenyl)propionate, Sumilizer BP 76, Tinogard TS, Tominokusu SS, Tominox SS, U 276, Ultranox 276, n-Octadecyl 3,5-di-tert-butyl-4-hydroxyhydrocinnamate, n-Octadecyl 3-(3,5-di-tert-butyl-4-hydroxyphenyl)propionate, n-Octadecyl 3-(4'-hydroxy-3',5'-di-tert-butylphenyl)propionate, n-Octadecyl β-(3,5-di-tert-butyl-4-hydroxyphenyl)propionate, n-Octadecyl β-(4'-hydroxy-3',5'-di-tert-butylphenyl)propionate, n-Octadecyl-3-(3',5'-di-tert-butyl-4'-hydroxyphenyl)propionate |
| InChI Formula | InChI=1S/C35H62O3/c1-8-9-10-11-12-13-14-15-16-17-18-19-20-21-22-23-26-38-32(36)25-24-29-27-30(34(2,3)4)33(37)31(28-29)35(5,6)7/h27-28,37H,8-26H2,1-7H3 |
| SMILES | CCCCCCCCCCCCCCCCCCOC(=O)CCc1cc(c(O)c(c1)C(C)(C)C)C(C)(C)C |
| IUPAC Name | octadecyl 3-(3,5-ditert-butyl-4-hydroxyphenyl)propanoate |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?