Learn More
CELLPATH OCT EMBEDDING MATRIX
Product Code. 30226281
Change view
Click to view available options
Form:
Liquid
Unit Size:
125mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Form Unit Size
30226281 Liquid 125mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30226281 Supplier CELLPATH Supplier No. KMA010000A
  • ONLINE EXCLUSIVE PRICE
    32.78 EUR
    12.38 EUR / 125mL
    Save 20.40 EUR
    62% Off
 Restricted Item
In Stock
Add to basket
Edge
Add to basket
OCT cryo microtomy embedding medium is designed for use at -20°C to -30°C. The medium freezes rapidly with minimal damage from crystal formation and is an ideal support medium for cutting tissue sections.
  • Key Benefits:
  • Liquid level visible in easy to use dispenser
  • Miscible with water to facilitate easy removal of tissue from the block holder
  • Easily dissolved from the microscope slide before staining
  • High viscosity to ensure strong adhesion of the tissue to the holder allowing correct orientation of the specimen while the medium is liquid
  • Freezes rapidly with minimal damage by crystal formation
  • Can be used as an infiltrating media for tissues that tend to shatter on cutting, such as brain
Specifications
Product Type Embedding Media
Form Liquid