Learn More
Oakton™ Zero Oxygen Calibration Solution
Product Code. 15327690 Shop All Cole Parmer Products
Change view
Click to view available options
Quantity:
500 mL
Unit Size:
500mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15327690 500 mL 500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 15327690 Supplier Cole Parmer Supplier No. 0065300
  • ONLINE EXCLUSIVE PRICE
    24.15 EUR
    23.31 EUR / 500mL
    Save 0.84 EUR
    3% Off
 Restricted Item
Estimated Shipment: 21-12-2026