Learn More
Oakton™ Replacement Electrode Holder for Portable Conductivity Meters
Product Code. 11599254
Change view
Click to view available options
For Use With (Equipment):
Oakton Conductivity Meters
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) Unit Size
11599254 Oakton Conductivity Meters Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 11599254 Supplier Oakton™ Supplier No. 15X000700
  • ONLINE EXCLUSIVE PRICE
    5.70 EUR
    4.95 EUR / Pack of 1
    Save 0.75 EUR
    13% Off
Estimated Shipment: 23-11-2026
Accessory electrode holder for Oakton 0.0 to 100.0°C Conductivity Meters.
For Oakton™ 0.0 to 100.0°C Conductivity Meters.
TRUSTED_SUSTAINABILITY
Specifications
Product Type Replacement Electrode Holder
Temperature Range (Metric) 0.0°C to 100.0°C
For Use With (Equipment) Oakton Conductivity Meters