Learn More
Oakton Hygrothermograph Chart Recorder Paper, 7 Day, 2 Hour
Product Code. 11739909 Shop All Cole Parmer Products
Change view
Click to view available options
Temperature Range (Metric):
-10°C to +50°C
Unit Size:
Pack of 100
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Temperature Range (Metric) Unit Size
11739909 -10°C to +50°C Pack of 100
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 11739909 Supplier Oakton Supplier No. 0836841
Estimated Shipment: 18-01-2027
Replacement Hygrothermograph Paper for Oakton™ hygrothermograph chart recorders.
  • Varying temperature ranges and speeds.
  • Choose the paper compatible with your chart recorder and application needs.
Specifications
Product Type Chart Paper
For Use With (Application) Two or three-speed type
For Use With (Equipment) Oakton™ Hygrothermograph Chart Recorders
Quantity 100 Pk.
Temperature Range (English) 14°F to 122°F
Temperature Range (Metric) -10°C to +50°C