missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ O8 OGG1 Inhibitor
GREENER_CHOICE

Product Code. 18769936
Change view
Click to view available options
Quantity:
5 mg
25 mg
Unit Size:
25mg
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18769936 25 mg 25mg
18769046 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18769936 Supplier Tocris Bioscience™ Supplier No. 6236/25

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Potent and selective 8-oxoguanine DNA glycosylase 1 (OGG1) inhibitor (IC50 = 0.2 μM in a gel based assay)

O8 OGG1 Inhibitor is a potent and selective 8-oxoguanine DNA glycosylase 1 (OGG1) inhibitor (IC50 = 0.2 μM in a gel based assay). Exhibits > 200-fold selectivity for OGG1 over other DNA glycosylases, NEIL1 and NTH1.
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 3,4-Dichlorobenzo[b]thiophene-2-carbohydrazide
CAS 350997-39-6
Quantity 25 mg
Molecular Formula C9H6Cl2N2OS
Formula Weight 261.13
Purity 98%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.