missing translation for 'onlineSavingsMsg'
Learn More

o-Xylene, Sigma-Aldrich™

Product Code. 15859680
Change view
Click to view available options
Quantity:
2.5 L
10 L
Unit Size:
10 L
2.5 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15859680 2.5 L 2.5 L
15836490 10 L 10 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Regulatory: This product is subject to excise duty on mineral oils in accordance with European Directive 2003/96/EC.

We regret to inform you that this product is not available for the Canary Islands, Ceuta and Melilla. We apologise for the inconvenience.
Product Code. 15859680 Supplier Sigma Aldrich Supplier No. 8086972500
 Restricted Item
Estimated Shipment: 27-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
Regulatory: This product is subject to excise duty on mineral oils in accordance with European Directive 2003/96/EC.

We regret to inform you that this product is not available for the Canary Islands, Ceuta and Melilla. We apologise for the inconvenience.

For synthesis

O-Xyleneis an aromatic hydrocarbon composed of two methyl substituents attached to adjacent carbon atoms. It has extremely low solubility in water. It is used to produce plasticizers, dyes, and pigments. Petroleum cracking produces O-Xylenes, which are used to generate phthalic anhydride, a precursor to many products, drugs, and chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 95-47-6

Specifications

Melting Point -26°C to -23°C (lit.)
Density 0.879 g/mL at 20°C
Quantity 2.5 L
Molecular Formula C6H4(CH3)2
MDL Number MFCD00008519
Synonym 1,2-Dimethylbenzene, 1,2-Dimethylbenzene
Boiling Point 143°C to 145°C
Molecular Weight (g/mol) 106.17
Grade Research
Chemical Name or Material O-Xylene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.