Learn More
o-Toluenesulfonamide-d7, TRC
Product Code. 30456073
Change view
Click to view available options
Quantity:
2.5 mg
10 mg
25 mg
Unit Size:
10mg
2.5mg
25mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30456073 25 mg 25mg
30452879 2.5 mg 2.5mg
30453172 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30456073 Supplier Toronto Research Chemicals Supplier No. TRCT53588225MG
 Restricted Item
Estimated Shipment: 12-11-2026
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material o-Toluenesulfonamide-d7
Molecular Formula C7 D7 H2 N O2 S
Quantity 25 mg
InChI Formula InChI=1S/C7H9NO2S/c1-6-4-2-3-5-7(6)11(8,9)10/h2-5H,1H3,(H2,8,9,10)/i1D3,2D,3D,4D,5D
SMILES [2H]c1c([2H])c([2H])c(c(c1[2H])C([2H])([2H])[2H])S(=O)(=O)N
IUPAC Name 2,3,4,5-tetradeuterio-6-(trideuteriomethyl)benzenesulfonamide
Molecular Weight (g/mol) 178.26
Formula Weight 178.079