missing translation for 'onlineSavingsMsg'
Learn More

o-Tolidine, Pract, MP Biomedicals

Product Code. 11429074 Shop All MP Biomedicals Products
Change view
Click to view available options
100g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
11429074 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11429074 Supplier MP Biomedicals Supplier No. 0521866480

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

CAS 119-93-7
Color Red to White
Density 1.0 (water = 1)
Molecular Formula C14H16N2
Quantity 100 g
Synonym o-tolidine, 3,3'-dimethylbenzidine, orthotolidine, diaminoditolyl, 2-tolidine, diaminotolyl, bianisidine, 3,3'-tolidine, 3,3'-dimethylbiphenyl-4,4'-diamine, 4,4'-bi-o-toluidine
InChI Key NUIURNJTPRWVAP-UHFFFAOYSA-N
SMILES CC1=C(C=CC(=C1)C2=CC(=C(C=C2)N)C)N
IUPAC Name 4-(4-amino-3-methylphenyl)-2-methylaniline
Molecular Weight (g/mol) 212.296
PubChem CID 8413
ChEBI CHEBI:34320
Formula Weight 212.3
Grade Practical
Physical Form Crystalline Powder or Crystals
Chemical Name or Material o-Tolidine
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.