Learn More
DWK Life Sciences O-ring, made of nitrile, for safety standard ground sockets
Shop All DWK Life Sciences ProductsDescription
Nitrile O-rings for DURAN™ safety ground joints together with the screw cap secure the connection between core and sleeve.
- In combination with the screw cap, the safety ground joints (core and sleeve) can be sealed tightly.
Specifications
Specifications
| For Use With (Application) | To secure the security grindings; Sealing |
| Joint Size | NS 29/32 |
| Quantity | 10/Pk. |
| Material | Nitrile |