Learn More
O,O,S-Trimethyl Ester Phosphorothioic Acid, TRC
Product Code. 30456337
Change view
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30456337 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30456337 Supplier Toronto Research Chemicals Supplier No. TRCT79589050MG
 Restricted Item
Estimated Shipment: 12-11-2026
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material O,O,S-Trimethylthiophosphate
CAS 152-20-5
Molecular Formula C3 H9 O3 P S
Quantity 50 mg
Synonym O,O,S-Trimethylphosphorthioate, [Methoxy(methylsulfanyl)phosphoryl]oxymethane
InChI Formula InChI=1S/C3H9O3PS/c1-5-7(4,6-2)8-3/h1-3H3
SMILES COP(=O)(OC)SC
IUPAC Name [methoxy(methylsulfanyl)phosphoryl]oxymethane
Molecular Weight (g/mol) 156.14
Formula Weight 156.001