missing translation for 'onlineSavingsMsg'
Learn More

(+)-O,O'-Dipivaloyl-D-tartaric acid, 98+%, Thermo Scientific™

Product Code. 15423368
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15423368 5 g 5g
15413368 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15423368 Supplier Thermo Scientific Alfa Aesar Supplier No. H56823.06

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 76769-55-6
Molecular Formula C14H22O8
Molecular Weight (g/mol) 318.32
MDL Number MFCD00015634
InChI Key UFHJEZDFEHUYCR-YZYOREDDNA-N
PubChem CID 16212279
SMILES CC(C)(C)C(=O)O[C@@H]([C@H](OC(=O)C(C)(C)C)C(O)=O)C(O)=O

Specifications

Melting Point 124°C to 126°C
Quantity 5 g
Beilstein 6894779
Sensitivity Hygroscopic
Synonym +-dipivaloyl-d-tartaric acid, 2s,3s-2,3-bis pivaloyloxy succinic acid, d-+-dipivaloyl tartaric acid, pubchem8173, --bis 2,2-dimethylpropionyl-l-tartaric acid, dipivaloyl-l-tartaric acid, +-dipivaloyl-d-tartaricacid, +-dpta, ?-o,o'-di-pivaloyl-l-tartaric acid, +-o,o'-di-pivaloyl-d-tartaric acid
Optical Rotation +24° (c=1.7 in Dioxane)
IUPAC Name (2S,3S)-2,3-bis(2,2-dimethylpropanoyloxy)butanedioic acid
Formula Weight 318.32
Percent Purity ≥98%
Chemical Name or Material (+)-O,O'-Dipivaloyl-D-tartaric acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.