missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ O-(N-Succinimidyl)-N,N,N',N'-bis(tetramethylene)uronium hexafluorophosphate, 98+%

Product Code. 15475265
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15475265 5 g 5g
15465265 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15475265 Supplier Alfa Aesar™ Supplier No. H54371.06

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 207683-26-9
Molecular Formula C13H20F6N3O3P
Molecular Weight (g/mol) 411.29
MDL Number MFCD00192457
InChI Key TVWKVGNCIJUXMY-UHFFFAOYSA-N
Synonym hspyu, dipyrrolidino n-succinimidyloxy carbenium hexafluorophosphate, n,n,n',n'-bis tetramethylene-o-n-succinimidyl uronium hexafluorophosphate, 1-2,5-dioxopyrrolidin-1-yl oxy pyrrolidin-1-yl methylene pyrrolidin-1-ium hexafluorophosphate v, 2,5-dioxopyrrolidin-1-yl oxy bis pyrrolidin-1-yl methylium hexafluorophosphate, dipyrrolidino n-succinimidyloxy carbenium hexafluorophosphate chn, bis pyrrolidin-1-yl methylidene 2,5-dioxopyrrolidin-1-yl oxidanium hexafluorophosphate, 1-1-pyrrolidin-1-iumylidene 1-pyrrolidinyl methoxy pyrrolidine-2,5-dione hexafluorophosphate, 1-pyrrolidin-1-ium-1-ylidene pyrrolidin-1-yl methoxy pyrrolidine-2,5-dione hexafluorophosphate, 1-2,5-dioxopyrrolidin-1-yl oxy pyrrolidin-1-yl methylidene-1??-pyrrolidin-1-ylium hexafluorophosphate
PubChem CID 16218611
IUPAC Name 1-{[(2,5-dioxopyrrolidin-1-yl)oxy](pyrrolidin-1-yl)methylidene}-1λ⁵-pyrrolidin-1-ylium; hexafluoro-λ⁵-phosphanuide
SMILES F[P-](F)(F)(F)(F)F.O=C1CCC(=O)N1OC(N1CCCC1)=[N+]1CCCC1

Specifications

Melting Point 152°C to 154°C
Quantity 5 g
Formula Weight 411.28
Percent Purity ≥98%
Chemical Name or Material O-(N-Succinimidyl)-N,N,N',N'-bis(tetramethylene)uronium hexafluorophosphate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.