missing translation for 'onlineSavingsMsg'
Learn More

O-Methyl Atorvastatin Calcium Salt, TRC

Product Code. 30012701
Change view
Click to view available options
Quantity:
2.5 mg
25 mg
Unit Size:
2.5mg
25mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30012701 25 mg 25mg
30007997 2.5 mg 2.5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30012701 Supplier Toronto Research Chemicals Supplier No. TRCM28827025MG
 Restricted Item
Estimated Shipment: 27-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 887196-29-4
Molecular Formula 2 C34 H36 F N2 O5 . Ca
Molecular Weight (g/mol) 1183.39
Synonym 1H-Pyrrole-1-heptanoic acid, 2-(4-fluorophenyl)-δ-hydroxy-β-methoxy-5-(1-methylethyl)-3-phenyl-4-[(phenylamino)carbonyl]-, calcium salt (2:1), (βR,δR)-, Calcium (βR,δR)-2-(4-fluorophenyl)-δ-hydroxy-β-methoxy-5-(1-methylethyl)-3-phenyl-4-[(phenylamino)carbonyl]-1H-pyrrole-1-heptanoate (1:2), (3R,5R)-7-[2-(4-Fluorophenyl)-5-(1-methylethyl)-3-phenyl-4-(phenylcarbamoyl)-1H-pyrrol-1-yl]-5-hydroxy-3-methoxyheptanoic acid calcium salt, 3-O-Methylatorvastatin calcium salt
IUPAC Name calcium;(3R,5R)-7-[2-(4-fluorophenyl)-3-phenyl-4-(phenylcarbamoyl)-5-propan-2-ylpyrrol-1-yl]-5-hydroxy-3-methoxyheptanoate
SMILES [Ca+2].CO[C@H](C[C@H](O)CCn1c(C(C)C)c(C(=O)Nc2ccccc2)c(c3ccccc3)c1c4ccc(F)cc4)CC(=O)[O-].CO[C@H](C[C@H](O)CCn5c(C(C)C)c(C(=O)Nc6ccccc6)c(c7ccccc7)c5c8ccc(F)cc8)CC(=O)[O-]

Specifications

Quantity 25 mg
InChI Formula InChI=1S/2C34H37FN2O5.Ca/c2*1-22(2)32-31(34(41)36-26-12-8-5-9-13-26)30(23-10-6-4-7-11-23)33(24-14-16-25(35)17-15-24)37(32)19-18-27(38)20-28(42-3)21-29(39)40;/h2*4-17,22,27-28,38H,18-21H2,1-3H3,(H,36,41)(H,39,40);/q;;+2/p-2/t2*27-,28-;/m11./s1
Formula Weight 1182.4842
Percent Purity >95%
Chemical Name or Material (3R,5R)-7-[2-(4-Fluorophenyl)-5-(1-methylethyl)-3-phenyl-4-(phenylcarbamoyl)-1H-pyrrol-1-yl]-5-hydroxy-3-methoxyheptanoic Acid Calcium Salt (3-O-Methylatorvastatin Calcium Salt)
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.