Learn More
O-Acetyl Isovanillin, TRC
Specifications
Specifications
| Chemical Name or Material | O-Acetyl Isovanillin |
| CAS | 881-57-2 |
| Molecular Formula | C10H10O4 |
| Quantity | 2.5 g |
| Synonym | 3-(Acetyloxy)-4-methoxy-benzaldehyde, 3-Hydroxy-p-anisaldehyde Acetate, 3-Acetoxy-4-methoxybenzaldehyde, 5-Formyl-2-methoxyphenyl Acetate |
| InChI Formula | InChI=1S/C10H10O4/c1-7(12)14-10-5-8(6-11)3-4-9(10)13-2/h3-6H,1-2H3 |
| SMILES | COc1ccc(C=O)cc1OC(=O)C |
| IUPAC Name | (5-formyl-2-methoxyphenyl) acetate |
| Molecular Weight (g/mol) | 194.18 |
| Formula Weight | 194.0579 |