Learn More
O-(4-Methoxybenzyl)hydroxylamine hydrochloride, 98%
Product Code. 15468236
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15468236 5 g 5g
15458236 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 15468236 Supplier Thermo Scientific Alfa Aesar Supplier No. H63312.06
Request Bulk or Custom Format
CAS: 876-33-5 | C8H12ClNO2 | 189.64 g/mol
O-(4-Methoxybenzyl)hydroxylamine hydrochloride is employed in intermediate of pharmaceutical, chemical research and analysis.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
O-(4-Methoxybenzyl)hydroxylamine hydrochloride is employed in intermediate of pharmaceutical, chemical research and analysis.
Notes
Store in cool, dry place in tightly closed container. With adequate ventilation. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 876-33-5
MDL Number MFCD01114582
Synonym o-4-methoxybenzyl hydroxylamine, o-4-methoxyphenyl methyl hydroxylamine, p-methoxybenzyloxyamine, hydroxylamine, o-p-methoxybenzyl, hydroxylamine, o-4-methoxyphenyl methyl, o-p-methoxybenzyl hydroxylamine, o-p-methoxybenzyl-hydroxylamine, o-4-methoxy-benzyl-hydroxylamine, 1-aminooxy methyl-4-methoxybenzene, 1-aminooxy methyl 4-methoxy-benzene
PubChem CID 120426
IUPAC Name O-[(4-methoxyphenyl)methyl]hydroxylamine
Specifications
Color White
Molecular Formula C8H12ClNO2
Quantity 5 g
InChI Key DHEZQYZJFCIQQA-UHFFFAOYSA-N
SMILES Cl.COC1=CC=C(CON)C=C1
Molecular Weight (g/mol) 189.64
Formula Weight 153.18
Percent Purity 98%
Physical Form Solid
Chemical Name or Material O-(4-Methoxybenzyl)hydroxylamine hydrochloride
compliance-icons
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H302-Harmful if swallowed.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.