Learn More
Masterflex™ Nylon Barb Adapter Tubing Connector
Product Code. 12447250 Shop All Masterflex Products
Change view
Click to view available options
Material:
Nylon
Unit Size:
Pack of 10
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Material Unit Size
12447250 Nylon Pack of 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 12447250 Supplier Masterflex™ Supplier No. 3061510
  • ONLINE EXCLUSIVE PRICE
    25.05 EUR
    23.61 EUR / Pack of 10
    Save 1.44 EUR
    5% Off
Estimated Shipment: 13-11-2026
Specifications
Material Nylon
Min. Operating Temperature (Metric) -46°C
Max. Operating Temperature (Metric) 121°C
Product Type Barb Adapter Tubing Connector