missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ NVS PAK1 C

Click to view available options
Quantity:
5 mg
25 mg
Unit Size:
25mg
5mg
Description
NVS PAK1 C is a negative control of NVS PAK1 1 (Cat. No. 6132), a potent and selective PAK1 inhibitor.
Specifications
Specifications
| Synonym | (3S)-3-(2-Chloro-5-(2,2-difluoroethyl)-8-fluoro-5H-dibenzo[b,e][1,4]diazepin-11-ylamino)-N,N-dimethylpyrrolidine-1-carboxamide |
| Quantity | 5 mg |
| Molecular Formula | C22H23ClF3N5O |
| Formula Weight | 465.91 |
| Purity | 98% |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction