missing translation for 'onlineSavingsMsg'
Learn More

NVP-ADW742, MedChemExpress

Product Code. 16486467
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16486467 5 mg 5mg
16483738 10 mg 10mg
16444806 100 mg 100mg
16420127 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16486467 Supplier Medchem Express Supplier No. HY10252
 Restricted Item
Estimated Shipment: 02-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress NVP-ADW742 (ADW742) is an orally active, selective IGF-1R tyrosine kinase inhibitor with an IC50 of 0.17 μM. NVP-ADW742 inhibits insulin receptor (InsR) with an IC50 of 2.8 μM. NVP-ADW742 induces pleiotropic antiproliferative/proapoptotic biologic sequelae in tumor cells.

NVP-ADW742 (ADW742) is an orally active, selective IGF-1R tyrosine kinase inhibitor with an IC50 of 0.17 μM. NVP-ADW742 inhibits insulin receptor (InsR) with an IC50 of 2.8 μM. NVP-ADW742 induces pleiotropic antiproliferative/proapoptotic biologic sequelae in tumor cells.

Specifications

Chemical Name or Material NVP-ADW742
CAS 475488-23-4
Color Light Yellow
Purity Grade Notes Research
Molecular Formula C28H31N5O
Quantity 5 mg
Synonym ADW742 GSK 552602A ADW
Solubility Information DMSO : 19.23 mg/mL (42.40 mM; Need ultrasonic)
SMILES NC1=C2C(N(C=C2C3=CC=CC(OCC4=CC=CC=C4)=C3)[C@H]5C[C@@H](C5)CN6CCCC6)=NC=N1
Molecular Weight (g/mol) 453.58
Formula Weight 453.58
Percent Purity 98.31%
Grade Research
Physical Form Powder
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.