Learn More
Syngene NuGenius XE
Product Code. 17922952 Shop All Syngene Products
Change view
Click to view available options
Pixel Size:
5 MP
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Pixel Size Unit Size
17922952 5 MP Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 17922952 Supplier Syngene Supplier No. NuGeniusXE
  • ONLINE EXCLUSIVE PRICE
    11028.00 EUR
    10400.49 EUR / Pack of 1
    Save 627.51 EUR
    5% Off
View all promotions
Estimated Shipment: 30-11-2026
NuGenius XE offers flexible DNA, stain free, autorads analysis. Intuitive touch screen system. Compact benchtop size with built in PC.
  • NuGenius XE offers flexible DNA, stain free, autorads analysis.
  • Intuitive touch screen system.
  • Compact benchtop size with built in PC.
  • Internal lighting options includes a UV transilluminator (purchased separately) for working with DNA gels (Ethidium bromide).
  • Our new UV-blue light converter screen (purchased separately) allows imaging of a wide range of safe dyes.
  • A visible converter option (purchased separately) can quickly extend its use for working with visible gels and blots (Coomassie blue and Silver stain).
  • The overhead LED white lighting is included as standard for easy sample positioning and focussing.
  • Safely excise your DNA bands from an agarose gel with the darkroom door open using the optional safety shield protecting the user when working with UV.
  • With a 5M pixel camera and f/1.2 motor driven lens provides exquisite pixel resolution and unrivalled sensitivity for gels and blots with a viewing area of 20 x 24 cm.
  • Images are saved directly to USB or network for easy transfer.
  • Quickly generate quantitative data with GeneTools analysis software.
  • NuGenius XE comes with unlimited copies of GeneTools.
Specifications
Pixel Size 5 MP
For Use With (Application) Gel Documentation
Product Type Gel Imaging System
Display Type Touchscreen