Learn More
Macherey-Nagel™ NucleoBond™ Smart Rack
Product Code. 15839097
Change view
Click to view available options
Quantity:
1 Ea.
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15839097 1 Ea. Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 15839097 Supplier Macherey-Nagel™ Supplier No. 740413
  • ONLINE EXCLUSIVE PRICE
    72.70 EUR
    59.33 EUR / Pack of 1
    Save 13.37 EUR
    18% Off
In Stock
Light grey rack for holding NucleoBond™ Xtra (Midi, Maxi, BAC) and NucleoBond™ AX (20, 100, 500, 2000, 10000) gravity flow columns. Rack holds up to six (6) Midi and four (4) Maxi columns during purification of plasmid DNA using anion exchange chromatography.
NucleoBond™ Xtra — Anion exchangers featuring fast filtration, high binding capacity and high flow rate.
  • Yields ultra-pure plasmid DNA in about half the time of other Midi or Maxi kits
  • NucleoBond™ Xtra Midi and Maxi kits contain enlarged columns creating shorter silica resin beds
  • Xtra Smart Rack hold up to 4 Maxi and 6 Midi columns for simultaneous processing
    • Columns are held by the collar ring of the column
    • Height of the column can be individually adjusted using plastic washers (included in the kit)
    • NucleoBond™ Xtra Smart Rack is sold separately from the NucleoBond™ Xtra kit
Specifications
Product Type Column holder/rack
For Use With (Application) Plasmid DNA purification
For Use With (Equipment) NucleBond™ Xtra Midi or Maxi kits
Quantity 1 Ea.
Samples per Run Up to 10