Learn More
NTB-A/SLAMF6/CD352 Antibody (005), PerCP, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand:  Novus Biologicals NBP2-90027PCP
520.20 EUR valid until 2025-12-16
BEST PRICE on Promo! Use promo code "24090" to get your promotional price.
Product Code. 18617240
  • 652.00 EUR / 0.01mL
Define Immunology Research on Your Terms With R&D Systems
Use promo code 24090
View all promotions
Description
NTB-A/SLAMF6/CD352 Monoclonal antibody specifically detects NTB-A/SLAMF6/CD352 in Human samples. It is validated for Immunohistochemistry (Paraffin)