missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ NSC 668036

Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
Description
NSC 668036 is a Wnt signaling inhibitor. Binds the PDZ domain of the Dishevelled (Dvl) protein. Inhibits proliferation, migration and TGF-β-induced differentiation of fibroblasts in vitro. Reduces severity of pulmonary fibrosis in a mouse model.
Specifications
Specifications
| Chemical Name or Material | N-[(1,1-dimethylethoxy)carbonyl]-L-alanyl-(2S)-2-hydroxy-3-methylbutanoyl-L-alanine-(1S)-1-carboxy-2-methylpropyl ester |
| CAS | 144678-63-7 |
| Quantity | 50 mg |
| Solubility | Soluble to 100mM in DMSO and to 100mM in ethanol |
| Molecular Formula | C21H36N2O9 |
| Formula Weight | 460.52 |
| Physical Form | Solid |
| Color | White |
| Product Type | Bioactive Small Molecule |
| Purity | >98% |
For Research Use Only.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction