missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NSC 601980 (analog), MedChemExpress
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
Description
NSC 601980 analog is the analog of the NSC601980, NSC601980 shows antitumor activity in the yeast screening experiment, which can inhibit cell proliferation in the COLO 205 and HT29 with Log GI 50 of -6.6 and -6.9 respectively.
Specifications
Specifications
| Chemical Name or Material | NSC 601980 (analog) |
| CAS | 91757-46-9 |
| Color | Yellow |
| Purity Grade Notes | Research |
| Molecular Formula | C15H14N4 |
| Quantity | 100 mg |
| Solubility Information | DMSO : ≥ 30 mg/mL (119.86 mM) |
| SMILES | CC1=NC2=CC=CC=C2N/C1=N/NC3=CC=CC=C3 |
| Molecular Weight (g/mol) | 250.3 |
| Formula Weight | 250.3 |
| Show More |
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction