Learn More
Invitrogen™ Blotting Tweezer
Click to view available options
Quantity:
1 unit
Unit Size:
Each
Description
Gasket fits on the cathode core (-) of the blot module. Tweezers designed for handling nitrocellulose or PVDF membranes.
1D Gel Electrophoresis, Protein Gel Electrophoresis, Proteins, Expression, Isolation and Analysis, Western Blotting
Specifications
Specifications
| For Use With (Application) | Protein assays and analysis, protein gel electrophoresis |
| For Use With (Equipment) | Bolt Mini Blot Module |
| Gel Size | Mini |
| Type | Tweezers |
| Quantity | 1 unit |