Learn More
Micronova™ NovaClean™ Lab and Glass Cleaner
BIOPHARMA
Product Code. 11962475 Shop All Micronova Products
Change view
Click to view available options
Volume (English):
1 qt.
1 gal.
Unit Size:
Pack of 4
Pack of 8
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume (English) Unit Size
11962475 1 gal. Pack of 4
11966855 1 qt. Pack of 8
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11962475 Supplier Micronova™ Supplier No. NC2G
  • ONLINE EXCLUSIVE PRICE
    302.00 EUR
    279.49 EUR / Pack of 4
    Save 22.51 EUR
    7% Off
 Restricted Item
Estimated Shipment: 14-01-2027
Ready to use, low-ionic detergent can be applied directly to a variety of cleanroom work surfaces including glass, stainless steel, epoxy acrylic, and vinyl curtains.
Lab and glass cleaner is a low-ionic detergent, designed for use on cleanroom work-stations, laminar flow benches, acrylic and painted surfaces.
  • Ready to use, cleanroom surfactant
  • Filtered to 0.1 microns
  • Low mobile ions
  • Low non-volatile residue levels
  • Can be applied directly to a range of surfaces
  • Works well with glass, stainless steel, epoxy, acrylic, and vinyl curtains
  • Ideal for conductive flooring
  • Available in a 1 quart spray bottle and a 1 gallon refill bottle
  • Biodegradable
  • Free-rinsing
Specifications
Product Type Filtered Low Ionic Surfactant (0.1 μ)
Color-coded No
Form Liquid
Handle Type None
pH 7.1
Product Line NovaClean
Scent Unscented
Sterility Non-sterile
Appearance Clear
Autoclavable Not Autoclavable
Boiling Point 99°C (210.2°F)
Care Instructions Avoid breathing vapor or mist and provide adequete ventilation when using.
Certifications/Compliance ISO 9001 Certified Facility; Certificate of Analysis
Color Clear
Container Type 1 Gallon Bottles
Content And Storage Store at ambient temperature. Avoid direct sunlight. Store in original container and keep tightly closed and seaqled until ready for use.
Density 1.025 g/mL at 25°C
Dimensions (L x W x H) 6 x 6 x 12 in.
Disposable Reusable
Filtered Filtered
Flammable No
For Use With (Application) Ready to use filtered cleanroom detergent, no mobile ions, free-rinsing for cleaning glass, stainless steel, acrylic and painted surfaces.
For Use With (Equipment) Mops and Buckets or Wipes and Sponge
Height (English) 12 in.
Height (Metric) 30.48 cm
Length (English) 6 in.
Length (Metric) 15.24 cm
Melting Point 0°C (32°F)
Packaging 4 bottles per poly-lined case
Quantity 4/Pk.
Resistant To The product is chemically stable. Under normal conditions of storage and use, hazardous reactions will not occur.
Size 13 x 13 x 13 in.
Vapor Pressure 2.3 kPa (17.1 mmHg)
Volume (English) 1 gal.
Volume (Metric) 3.79 L
Width (English) 6 in.
Width (Metric) 15.24 cm