missing translation for 'onlineSavingsMsg'
Learn More

Notoginsenoside R2, MedChemExpress

Product Code. 16431598
Encompass_Preferred
Click to view available options
Quantity:
5 mg
Unit Size:
5mg
This item is not returnable. View return policy

Product Code. 16431598

Brand: Medchem Express HYN0909

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Notoginsenoside R2 is a newly isolated notoginsenoside from Panax notoginseng, showed neuroprotective effects against 6-OHDA-induced oxidative stress and apoptosis.

Notoginsenoside R2 is a newly isolated notoginsenoside from Panax notoginseng, showed neuroprotective effects against 6-OHDA-induced oxidative stress and apoptosis.

Specifications

Chemical Name or Material Notoginsenoside R2
CAS 80418-25-3
Color White
Purity Grade Notes Research
Molecular Formula C41H70O13
Quantity 5 mg
Synonym 20(S)-Notoginsenoside R2 Ginsenoside Ng-R2
Solubility Information DMSO : 100 mg/mL (129.70 mM; Need ultrasonic)
SMILES C[C@]([C@@](C[C@H]1O)([H])[C@]2(CC[C@@H]3O)C)(C[C@H](O[C@@](O[C@H](CO)[C@@H](O)[C@@H]4O)([H])[C@@H]4O[C@@](OC[C@@H](O)[C@@H]5O)([H])[C@@H]5O)[C@@]2([H])C3(C)C)[C@]6([C@@]1([H])[C@]([C@@](C)(O)CC/C=C(C)/C)([H])CC6)C
Molecular Weight (g/mol) 770.99
Formula Weight 770.99
Percent Purity 98.14%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neuromodulation
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.