missing translation for 'onlineSavingsMsg'
Learn More

Norneo Vardenafil, TRC

Product Code. 30137609
Change view
Click to view available options
Quantity:
100 mg
1 g
Unit Size:
100mg
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30137609 1 g 1g
17934615 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30137609 Supplier Toronto Research Chemicals Supplier No. TRCN8245001G
 Restricted Item
Estimated Shipment: 24-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 358390-39-3
Molecular Formula C18 H20 N4 O4
Molecular Weight (g/mol) 356.38
Synonym 3-(1,4-Dihydro-5-methyl-4-oxo-7-propylimidazo[5,1-f][1,2,4]triazin-2-yl)-4-ethoxybenzoic acid, Norneovardenafil
IUPAC Name 4-ethoxy-3-(5-methyl-4-oxo-7-propyl-3H-imidazo[5,1-f][1,2,4]triazin-2-yl)benzoic acid
SMILES CCCc1nc(C)c2C(=O)NC(=Nn12)c3cc(ccc3OCC)C(=O)O

Specifications

Quantity 1 g
InChI Formula InChI=1S/C18H20N4O4/c1-4-6-14-19-10(3)15-17(23)20-16(21-22(14)15)12-9-11(18(24)25)7-8-13(12)26-5-2/h7-9H,4-6H2,1-3H3,(H,24,25)(H,20,21,23)
Formula Weight 356.1485
Percent Purity >95%
Chemical Name or Material Norneo Vardenafil
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.