missing translation for 'onlineSavingsMsg'
Learn More

Norflurazon, TRC

Product Code. 30419469
Change view
Click to view available options
Quantity:
50 mg
100 mg
1 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30419469 1 g
30410351 50 mg
30418510 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30419469 Supplier Toronto Research Chemicals Supplier No. TRCN6845001G
 Restricted Item
Estimated Shipment: 27-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 27314-13-2
Molecular Formula C12 H9 Cl F3 N3 O
Molecular Weight (g/mol) 303.67
Synonym Evital, 4-chloro-5-(methylamino)-2-(α,α,α-trifluoro-m-tolyl)pyridazin-3(2H)-one
IUPAC Name 4-chloro-5-(methylamino)-2-[3-(trifluoromethyl)phenyl]pyridazin-3-one
SMILES CNC1=C(Cl)C(=O)N(N=C1)c2cccc(c2)C(F)(F)F

Specifications

Quantity 1 g
InChI Formula InChI=1S/C12H9ClF3N3O/c1-17-9-6-18-19(11(20)10(9)13)8-4-2-3-7(5-8)12(14,15)16/h2-6,17H,1H3
Formula Weight 303.0386
Percent Purity >95%
Chemical Name or Material Norflurazon
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.