Description
Honeywell Fluka™ offers an expanding range of high quality inorganic certified reference materials and organic reference materials.
- Produced according to ISO 9001
- Suitable for a wide range of industries and applications
- Detailed Certificate of Analysis included with every delivery
Specifications
Specifications
| Chemical Name or Material | Norfloxacin-d5 |
| CAS | 1015856-57-1 |
| CAS Max % | <100% |
| Color | White |
| Melting Point | 220°C to 223°C |
| Synonym | 6-Fluoro-1,4-dihydro-4-oxo-1-pentadeuteroethyl-7-piperazino-3-quinolinecarboxylic acid |
| Molecular Weight (g/mol) | 324.36 g/mol |
| Quantity | 10 mg |
| Grade | Antibiotics |