missing translation for 'onlineSavingsMsg'
Learn More

Nordihydorguaiaretic acid, 99%, For HPLC analysis, MP Biomedicals™

Product Code. 11459231 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11459231 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11459231 Supplier MP Biomedicals Supplier No. 0215594491
  • ONLINE EXCLUSIVE PRICE
    197.00 EUR
    186.90 EUR / 1g
    Save 10.10 EUR
    5% Off
 Restricted Item
Estimated Shipment: 21-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 500-38-9
Color White
Molecular Formula C18H22O4
Quantity 1 g
Synonym nordihydroguaiaretic acid, ndga, dihydronorguaiaretic acid, norhydroguaiaretic acid, nordihydroguairaretic acid, norguaiaretic acid, dihydro, 1,2-benzenediol, 4,4'-2,3-dimethyl-1,4-butanediyl bis, dinorguaiaretic acid, dihydro, 4,4'-2,3-dimethyltetramethylene dipyrocatechol, 2,3-bis 3,4-dihydroxyphenylmethyl butane
InChI Key HCZKYJDFEPMADG-UHFFFAOYSA-N
SMILES CC(CC1=CC(=C(C=C1)O)O)C(C)CC2=CC(=C(C=C2)O)O
IUPAC Name 4-[4-(3,4-dihydroxyphenyl)-2,3-dimethylbutyl]benzene-1,2-diol
Molecular Weight (g/mol) 302.37
PubChem CID 4534
ChEBI CHEBI:7625
Formula Weight 302.4
Percent Purity 0.99
Physical Form Crystalline Powder
Chemical Name or Material Nordihydorguaiaretic acid, For HPLC analysis
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.