missing translation for 'onlineSavingsMsg'
Learn More

Nonanamide, TRC

Product Code. 30179960
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30179960 5 mg 5mg
30180141 50 mg 50mg
30180220 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30179960 Supplier Toronto Research Chemicals Supplier No. TRCB5043685MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1120-07-6
Molecular Formula C9H19NO
Molecular Weight (g/mol) 157.25
IUPAC Name nonanamide
SMILES CCCCCCCCC(=O)N

Specifications

Quantity 5 mg
InChI Formula InChI=1S/C9H19NO/c1-2-3-4-5-6-7-8-9(10)11/h2-8H2,1H3,(H2,10,11)
Formula Weight 157.1467
Chemical Name or Material Nonanamide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.