missing translation for 'onlineSavingsMsg'
Learn More

Nolatrexed dihydrochloride, MedChemExpress

Product Code. 16470547
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16470547 50 mg 50mg
16476917 5 mg 5mg
16414208 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 16470547 Supplier Medchem Express Supplier No. HY108474
 Restricted Item
Estimated Shipment: 05-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress Nolatrexed dihydrochloride (AG 337) is a non-competitive lipophilic inhibitor of thymidylate synthase, interacts at the folate cofactor binding site of the enzyme, with a Ki of 11 nM for human thymidylate synthase. Nolatrexed dihydrochloride (AG 337) induces cell cycle arrest in S phase of cancer cells. Anti-cancer activity.

Nolatrexed dihydrochloride (AG 337) is a non-competitive lipophilic inhibitor of thymidylate synthase, interacts at the folate cofactor binding site of the enzyme, with a Ki of 11 nM for human thymidylate synthase. Nolatrexed dihydrochloride (AG 337) induces cell cycle arrest in S phase of cancer cells. Anti-cancer activity.

Specifications

Chemical Name or Material Nolatrexed dihydrochloride
Physical Form Solid
CAS 152946-68-4
Color Yellow
Quantity 50 mg
Purity Grade Notes Research
Molecular Formula C14H14Cl2N4OS
For Use With (Application) Cancer-programmed cell death
Synonym AG 337 Thymitaq
Solubility Information H2O : 50 mg/mL (139.95 mM; Need ultrasonic) ∣DMSO : 41.67 mg/mL (116.64 mM; Need ultrasonic)
SMILES [H]Cl.O=C1N=C(N)NC2=C1C(SC3=CC=NC=C3)=C(C)C=C2.[H]Cl
Molecular Weight (g/mol) 357.26
Formula Weight 357.26
Percent Purity 98.54%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.