missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Nolatrexed dihydrochloride, MedChemExpress
Description
Nolatrexed dihydrochloride (AG 337) is a non-competitive lipophilic inhibitor of thymidylate synthase, interacts at the folate cofactor binding site of the enzyme, with a Ki of 11 nM for human thymidylate synthase. Nolatrexed dihydrochloride (AG 337) induces cell cycle arrest in S phase of cancer cells. Anti-cancer activity.
Specifications
Specifications
| Chemical Name or Material | Nolatrexed dihydrochloride |
| Physical Form | Solid |
| CAS | 152946-68-4 |
| Color | Yellow |
| Quantity | 5 mg |
| Purity Grade Notes | Research |
| Molecular Formula | C14H14Cl2N4OS |
| For Use With (Application) | Cancer-programmed cell death |
| Synonym | AG 337 Thymitaq |
| Solubility Information | H2O : 50 mg/mL (139.95 mM; Need ultrasonic) ∣DMSO : 41.67 mg/mL (116.64 mM; Need ultrasonic) |
| Show More |
Research purposes only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?