Learn More
Nizatidine EP Impurity H (as Hydrochloride), Mikromol™
Product Code. 30457639
Change view
Click to view available options
Quantity:
25 mg
100 mg
4 x 25 mg
Unit Size:
100mg
25mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30457639 4 x 25 mg -
30049587 25 mg 25mg
30050135 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30457639 Supplier Mikromol Supplier No. MM0220.1100254AMP
 Restricted Item
Estimated Shipment: 30-11-2026
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
Nizatidine EP Impurity H (as Hydrochloride), Mikromol
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material 2-(Dimethylamino)thioacetamide Hydrochloride
CAS 27366-72-9
Analyte or Component Names 2-(Dimethylamino)thioacetamide Hydrochloride
Shipping Condition Room Temperature
Molecular Formula C4 H10 N2 S . Cl H
Certifications/Compliance ISO 9001
Synonym 2-(Dimethylamino)thioacetamide Hydrochloride, Nizatidine Imp. H (EP) as Hydrochloride, Ethanethioamide, 2-(dimethylamino)-, hydrochloride (1:2), Nizatidine Impurity H as Hydrochloride
InChI Formula InChI=1S/C4H10N2S.ClH/c1-6(2)3-4(5)7;/h3H2,1-2H3,(H2,5,7);1H
Solution Type Neat
SMILES Cl.CN(C)CC(=S)N
IUPAC Name 2-(dimethylamino)ethanethioamide;hydrochloride
Molecular Weight (g/mol) 154.66
Quantity 4 x 25 mg
Formula Weight 154.0331 g/mol