Learn More
MedChemExpress Nivolumab
MedChemExpress Nivolumab is a programmed death receptor-1 (PD-1) blocking human IgG4 antibody to treat advanced (metastatic) non-small cell lung cancer.
Brand: MedChemExpress HY-P9903
Specifications
| PD-1/PD-L1 | |
| Please store the product under the recommended conditions in the Certificate of Analysis. | |
| Shipping with dry ice | |
| 98.29% |
| 5 mg | |
| Inhibitory Antibodies | |
| 946414-94-4 |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.