Learn More
Lenz Laborglasintrumente Nitrogen Tubes (Schlenk Tubes) with PTFE Needle-Valve Stopcock
Product Code. 11923421
Change view
Click to view available options
Capacity (Metric):
10 mL
250 mL
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) unitSize
11923421 250 mL Each
11983411 10 mL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11923421 Supplier Lenz Laborglasintrumente Supplier No. 03370649
  • ONLINE EXCLUSIVE PRICE
    124.00 EUR
    76.80 EUR / Pack of 1
    Save 47.20 EUR
    38% Off
Estimated Shipment: 10-09-2026
Add to basket
Add to basket
Made of high quality materials. Each manufacturing step is tested through an extensive quality management system, based on DIN ISO 9001/EN 29001.
  • PTFE needle-valve stopcock
  • With conical ground joint and PTFE stopcock
  • Made of DURAN™ tubing
Specifications
Capacity (Metric) 250 mL
Material Duran Glass