missing translation for 'onlineSavingsMsg'
Learn More

Nitrofurantoin, 99.5%, MP Biomedicals™

Product Code. 11449161 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
100 g
Unit Size:
100g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11449161 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11449161 Supplier MP Biomedicals Supplier No. 0215588180

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 67-20-9
Color Orange-Yellow to Yellow
Assay Percent Range 0.995
Molecular Formula C8H6N4O5
MDL Number MFCD00003224
Quantity 100 g
Synonym nitrofurantoin, macrodantin, 5-nitrofurantoin, furadantin, furadonine, nitrofurantoine, furadantine, furadantoin, furadoine, furadontin
InChI Key NXFQHRVNIOXGAQ-YCRREMRBSA-N
SMILES [O-][N+](=O)C1=CC=C(O1)\C=N\N1CC(=O)NC1=O
IUPAC Name 1-[(E)-[(5-nitrofuran-2-yl)methylidene]amino]imidazolidine-2,4-dione
Molecular Weight (g/mol) 238.16
PubChem CID 6604200
ChEBI CHEBI:71415
Formula Weight 238.2
Physical Form Crystalline Powder
Chemical Name or Material Nitrofurantoin
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.