missing translation for 'onlineSavingsMsg'
Learn More

Nitrilotriacetic Acid, ACS Grade, ≥98%, MP Biomedicals™

Product Code. 11442892 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
100 g
1 kg
Unit Size:
100g
1kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11442892 1 kg 1kg
11422892 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11442892 Supplier MP Biomedicals Supplier No. 0215255191
 Restricted Item
Estimated Shipment: 12-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 139-13-9
Molecular Formula C6H9NO6
Molecular Weight (g/mol) 191.14
MDL Number MFCD00004287
InChI Key MGFYIUFZLHCRTH-UHFFFAOYSA-N
Synonym nitrilotriacetic acid, 2,2',2-nitrilotriacetic acid, triglycollamic acid, aminotriacetic acid, complexon i, trilon a, n,n-bis carboxymethyl glycine, nitrilotriacetate, komplexon i, titriplex i
PubChem CID 8758
ChEBI CHEBI:44557
IUPAC Name 2-[bis(carboxymethyl)amino]acetic acid
SMILES OC(=O)CN(CC(O)=O)CC(O)=O

Specifications

Color White
Assay Percent Range ≥98%
Quantity 1 kg
Formula Weight 191.14
Grade ACS Reagent
Physical Form Solid
Chemical Name or Material Nitrilotriacetic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.