missing translation for 'onlineSavingsMsg'
Learn More

Nitrilotriacetic Acid, ACS Grade, ≥98%, MP Biomedicals™

Product Code. 11432892 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
500 g
Unit Size:
500g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11432892 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11432892 Supplier MP Biomedicals Supplier No. 0215255190
 Restricted Item
Estimated Shipment: 12-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 139-13-9
Assay Percent Range ≥98%
Molecular Formula C6H9NO6
MDL Number MFCD00004287
Quantity 500 g
Synonym Nitrilotriacetic acid, Aminotriacetic acid
InChI Key MGFYIUFZLHCRTH-UHFFFAOYSA-N
SMILES OC(=O)CN(CC(O)=O)CC(O)=O
IUPAC Name 2-[bis(carboxymethyl)amino]acetic acid
Molecular Weight (g/mol) 191.14
Formula Weight 191.14
Grade ACS Reagent
Physical Form Powder
Chemical Name or Material Nitrilotriacetic Acid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.