missing translation for 'onlineSavingsMsg'
Learn More

Nitric Acid (65%), TRC

Product Code. 30179758
Change view
Click to view available options
Quantity:
100 mL
250 mL
Unit Size:
100mL
250mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30179758 100 mL 100mL
30180400 250 mL 250mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Regulatory: This product contains an explosive precursor according to Regulation (EC) No 1148/2019, all customers must complete a declaration before delivery.
Product Code. 30179758 Supplier Toronto Research Chemicals Supplier No. TRCN481520100ML

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: This product contains an explosive precursor according to Regulation (EC) No 1148/2019, all customers must complete a declaration before delivery.

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7697-37-2
Molecular Formula H N O3
Molecular Weight (g/mol) 63.01
Synonym Nitric acid
IUPAC Name nitric acid
SMILES O[N+](=O)[O-]

Specifications

Quantity 100 mL
InChI Formula InChI=1S/HNO3/c2-1(3)4/h(H,2,3,4)
Formula Weight 62.9956
Chemical Name or Material Nitric acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.