missing translation for 'onlineSavingsMsg'
Learn More

Nitric Acid (65%), TRC

Product Code. 30482741
Change view
Click to view available options
Quantity:
25 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30482741 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30482741 Supplier Toronto Research Chemicals Supplier No. TRCN48152025ML

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7697-37-2
Molecular Formula H N O3
Molecular Weight (g/mol) 63.01
Synonym Nitric acid
IUPAC Name nitric acid
SMILES O[N+](=O)[O-]

Specifications

Concentration 0.65
Quantity 25 mL
InChI Formula InChI=1S/HNO3/c2-1(3)4/h(H,2,3,4)
Formula Weight 62.9956
Chemical Name or Material Nitric acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.