missing translation for 'onlineSavingsMsg'
Learn More

Nimorazole, MedChemExpress

Product Code. 16463087
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
200 mg
500 mg
Unit Size:
100mg
10mg
200mg
500mg
50mg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16463087 50 mg 50mg
16457038 10 mg 10mg
16451806 500 mg 500mg
16417096 100 mg 100mg
16401226 200 mg 200mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 16463087 Supplier Medchem Express Supplier No. HY16349
 Restricted Item
Estimated Shipment: 17-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress Nimorazole (K-1900), a 2-nitroimidazole, is a hypoxic cell-radiation sensitizer. Nimorazole has anti-infective and anti-protozoal against trichomoniasis. Nimorazole has the potential for head and neck cancer.

Nimorazole (K-1900), a 2-nitroimidazole, is a hypoxic cell-radiation sensitizer. Nimorazole has anti-infective and anti-protozoal against trichomoniasis. Nimorazole has the potential for head and neck cancer.

Specifications

Chemical Name or Material Nimorazole
Physical Form Powder
CAS 6506-37-2
Color Light Brown
Quantity 50 mg
Purity Grade Notes Research
Molecular Formula C9H14N4O3
For Use With (Application) COVID-19-immunoregulation
Synonym K-1900
Solubility Information DMSO : 33.33 mg/mL (147.33 mM; Need ultrasonic)
SMILES O=[N+](C1=CN=CN1CCN2CCOCC2)[O-]
Molecular Weight (g/mol) 226.23
Formula Weight 226.23
Percent Purity 98.05%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.