Learn More
Nifurtimox, MedChemExpress
Product Code. 16462698
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
Unit Size:
10mg
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16462698 5 mg 5mg
16410489 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 16462698 Supplier Medchem Express Supplier No. HYW040073
 Restricted Item
Estimated Shipment: 17-08-2026
Add to basket
Add to basket
MedChemExpress Nifurtimox, an antiprotozoal agent, which is generally used for the treatment of infections with Trypanosoma cruzi, has been used in the therapy of neuroblastoma. Nifurtimox affects enzyme activity of lactate dehydrogenase (LDH).
Nifurtimox, an antiprotozoal agent, which is generally used for the treatment of infections with Trypanosoma cruzi, has been used in the therapy of neuroblastoma. Nifurtimox affects enzyme activity of lactate dehydrogenase (LDH).
Specifications
Chemical Name or Material Nifurtimox
Physical Form Solid
CAS 23256-30-6
Color Yellow
Quantity 5 mg
Purity Grade Notes Research
Molecular Formula C10H13N3O5S
For Use With (Application) Cancer-programmed cell death
Solubility Information DMSO : 150 mg/mL (522.10 mM; Need ultrasonic)
SMILES O=S1(CC(C)N(/N=C/C2=CC=C([N+]([O-])=O)O2)CC1)=O
Molecular Weight (g/mol) 287.3
Formula Weight 287.3
Percent Purity 99.65%
Grade Research