missing translation for 'onlineSavingsMsg'
Learn More

Nifuroxazide, 99.9%, MP Biomedicals™

Product Code. 11415041 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
50 g
Unit Size:
50g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11415041 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11415041 Supplier MP Biomedicals Supplier No. 0215583350

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

CAS 98-95-3
Color Yellow
Assay Percent Range 0.999
Molecular Formula C6H5NO2
Quantity 50 g
Synonym nitrobenzol, benzene, nitro, oil of mirbane, mirbane oil, essence of mirbane, nitro-benzene, nitrobenzeen, oil of myrbane, nitrobenzen, mononitrobenzene
InChI Key LQNUZADURLCDLV-UHFFFAOYSA-N
SMILES C1=CC=C(C=C1)[N+](=O)[O-]
IUPAC Name nitrobenzene
Molecular Weight (g/mol) 123.111
PubChem CID 7416
ChEBI CHEBI:27798
Formula Weight 275.22
Physical Form Powder
Chemical Name or Material Nifuroxazide
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.