missing translation for 'onlineSavingsMsg'
Learn More

Nicotinoyl Chloride Hydrochloride, TRC

Product Code. 30470503
Change view
Click to view available options
Quantity:
25 g
Unit Size:
25g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30470503 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30470503 Supplier Toronto Research Chemicals Supplier No. TRCN43125025G
 Restricted Item
Estimated Shipment: 08-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Nicotinoyl Chloride Hydrochloride
CAS 20260-53-1
Molecular Formula C6H5Cl2NO
Quantity 25 g
Synonym 3-Pyridinecarbonyl Chloride Hydrochloride, Nicotinic Acid Chloride Hydrochloride
InChI Formula InChI=1S/C6H4ClNO.ClH/c7-6(9)5-2-1-3-8-4-5;/h1-4H;1H
SMILES Cl.ClC(=O)c1cccnc1
IUPAC Name pyridine-3-carbonyl chloride;hydrochloride
Molecular Weight (g/mol) 178.02
Formula Weight 176.9748
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.