missing translation for 'onlineSavingsMsg'
Learn More

Nicotinic Acid, Sigma-Aldrich™

Product Code. 15830581
Change view
Click to view available options
Quantity:
100 g
1 kg
Unit Size:
100g
1kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15830581 1 kg 1kg
15820581 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15830581 Supplier Sigma Aldrich Supplier No. 8187141000
 Restricted Item
Estimated Shipment: 16-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

For synthesis

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 59-67-6

Specifications

Melting Point 236.6°C
Density 1.47 g/cm3 at 20°C
Molecular Formula 3-(COOH)-C5H4N
MDL Number MFCD00006391
Quantity 1 kg
Synonym Nicotinic acid, Niacin, 3-Pyridinecarboxylic acid, Nicotinic acid
Molecular Weight (g/mol) 123.11
Grade Research
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.