missing translation for 'onlineSavingsMsg'
Learn More

Nicotinic Acid Diethylamide, 99.8%, MP Biomedicals™

Product Code. 11493700 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
25 mL
100 mL
Unit Size:
100mL
25mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11493700 25 mL 25mL
11403710 100 mL 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11493700 Supplier MP Biomedicals Supplier No. 0210244525

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 59-26-7
Molecular Formula C10H14N2O
Molecular Weight (g/mol) 178.235
InChI Key NCYVXEGFNDZQCU-UHFFFAOYSA-N
Synonym nikethamide, n,n-diethylnicotinamide, nicethamide, corvin, anacardone, anacordone, carbamidal, coramine, corvotone, nicamide
PubChem CID 5497
IUPAC Name N,N-diethylpyridine-3-carboxamide
SMILES CCN(CC)C(=O)C1=CN=CC=C1

Specifications

Color Colorless
Quantity 25 mL
Formula Weight 178.2
Physical Form Liquid
Chemical Name or Material Nicotinic Acid Diethylamide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.